2-Fluoro-4-hydroxybenzonitrile
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
AS24820G100
AS24820G25
ACTIAS24820G100EA
196
GBP
ACTIAS24820G100
ACTIAS24820G25
2-Fluoro-4-hydroxybenzonitrile
2-Fluoro-4-hydroxybenzonitrile
Formula:
C₇H₄FNO MW: 137.11 g/mol Boiling Pt: 261 °C (1013 hPa) Melting Pt: ∼124 °C Density: 1,45 g/cm³ (20 °C) |
MDL Number:
MFCD00192177 CAS Number: 82380-18-5 EINECS: 422-810-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...