2-Amino-3,5-dichlorobenzonitrile

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS21305G25 AS21305G5
ACTIAS21305G25EA 229 GBP
ACTIAS21305G25 ACTIAS21305G5
2-Amino-3,5-dichlorobenzonitrile
2-Amino-3,5-dichlorobenzonitrile
Formula: C₇H₄Cl₂N₂
MW: 187.03 g/mol
MDL Number: MFCD00158947
CAS Number: 36764-94-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR