2-Methoxynicotinic acid

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2-Methoxy-3-pyridinecarboxylic acid, 2-Methoxypyridine-3-carboxylic acid

AS21960G100 AS21960G25
ACTIAS21960G100EA 0 GBP
ACTIAS21960G100 ACTIAS21960G25
2-Methoxynicotinic acid
2-Methoxynicotinic acid
Formula: C₇H₇NO₃
MW: 153.14 g/mol
MDL Number: MFCD00085110
CAS Number: 16498-81-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR