tert-Butyl cyanoacetate
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
tert-Butyl 2-cyanoacetate
AS22706G100
AS22706G25
ACTIAS22706G100EA
0
GBP
ACTIAS22706G100
ACTIAS22706G25
tert-Butyl cyanoacetate
tert-Butyl cyanoacetate
Formula:
C₇H₁₁NO₂ MW: 141.17 g/mol Density: 0.987 g/cm³ (20 °C) |
MDL Number:
MFCD00001938 CAS Number: 1116-98-9 EINECS: 214-243-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...