5-Isoxazolecarboxylic acid

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Isoxazole-5-carboxylic acid

AS25294G1 AS25294G25 AS25294G5
ACTIAS25294G1EA 14 GBP
ACTIAS25294G1 ACTIAS25294G25 ACTIAS25294G5
5-Isoxazolecarboxylic acid
5-Isoxazolecarboxylic acid
Formula: C₄H₃NO₃
MW: 113.07 g/mol
MDL Number: MFCD00156151
CAS Number: 21169-71-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR