5-Bromothiazole

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS50158G1 AS50158G25 AS50158G5
ACTIAS50158G1EA 18 GBP
ACTIAS50158G1 ACTIAS50158G25 ACTIAS50158G5
5-Bromothiazole
5-Bromothiazole
Formula: C₃H₂BrNS
MW: 164.03 g/mol
MDL Number: MFCD07787394
CAS Number: 3034-55-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR