5-Amino-1H-indazole
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
1H-Indazol-5-amine, Indazol-5-amine, 5-Amino-1H-indazole, 5-Aminoindazole, 1H-Indazol-5-ylamine, Indazol-5-ylamine
AS20477G100
AS20477G25
AS20477G5
ACTIAS20477G100EA
292
GBP
ACTIAS20477G100
ACTIAS20477G25
ACTIAS20477G5
5-Amino-1H-indazole
5-Amino-1H-indazole
Formula:
C₇H₇N₃ MW: 133.15 g/mol Boiling Pt: 345 °C (1013 hPa) Melting Pt: 183 °C |
MDL Number:
MFCD00037975 CAS Number: 19335-11-6 EINECS: 242-971-5 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...