4-Iodopyridine

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS51458G25 AS51458G5
ACTIAS51458G25EA 0 GBP
ACTIAS51458G25 ACTIAS51458G5
4-Iodopyridine
4-Iodopyridine
Formula: C₅H₄IN
MW: 205 g/mol
MDL Number: MFCD02093937
CAS Number: 15854-87-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR