6-Chlorouracil
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
6-Chloro-2,4-dihydroxypyrimidine, 6-Chloropyrimidine-2,4(1H,3H)-dione, 4-Chlorouracil, 4-(3-Bromophenyl)pyrazole, 4-Chloro-2,6-dihydroxypyrimidine, 6-Chloro-2,4(1H,3H)-pyrimidinedione
AS23249G100
AS23249G25
ACTIAS23249G100EA
88
GBP
ACTIAS23249G100
ACTIAS23249G25
6-Chlorouracil
6-Chlorouracil
Formula:
C₄H₃ClN₂O₂ MW: 146.53 g/mol |
MDL Number:
MFCD00014595 CAS Number: 4270-27-3 EINECS: 224-258-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...