4-Bromopyridinium chloride

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: 4-Bromopyridine HCl, 4-Bromopyridine hydrochloride

AS50279G100 AS50279G25
ACTIAS50279G100EA 0 GBP
ACTIAS50279G100 ACTIAS50279G25
4-Bromopyridinium chloride
4-Bromopyridinium chloride
Formula: C₅H₅BrClN
MW: 194.46 g/mol
Melting Pt: 235…238 °C
MDL Number: MFCD00012828
CAS Number: 19524-06-2
EINECS: 243-128-4
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR