4-(1-Aminoethyl)pyridine

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 1-Pyridin-4-ylethylamine, 1-(4-Pyridyl)ethylamine

AS21928G1 AS21928G25 AS21928G5
ACTIAS21928G1EA 43 GBP
ACTIAS21928G1 ACTIAS21928G25 ACTIAS21928G5
4-(1-Aminoethyl)pyridine
4-(1-Aminoethyl)pyridine
Formula: C₇H₁₀N₂
MW: 122.17 g/mol
MDL Number: MFCD02245004
CAS Number: 50392-78-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR