3-Thenoic acid

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 3-Thiophenecarboxylic acid, Thiophene-3-carboxylic acid

AS20110G100 AS20110G25
ACTIAS20110G100EA 0 GBP
ACTIAS20110G100 ACTIAS20110G25
3-Thenoic acid
3-Thenoic acid
Formula: C₅H₄O₂S
MW: 128.15 g/mol
Melting Pt: 137 °C
MDL Number: MFCD00005467
CAS Number: 88-13-1
EINECS: 201-802-5
Merck Index: 12,09415

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR