2-Bromothiazole

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS50234G100 AS50234G25
ACTIAS50234G100EA 0 GBP
ACTIAS50234G100 ACTIAS50234G25
2-Bromothiazole
2-Bromothiazole
Formula: C₃H₂BrNS
MW: 164.03 g/mol
Boiling Pt: 171 °C (1013 hPa)
Density: 1,82 g/cm³ (20 °C)
MDL Number: MFCD00005316
CAS Number: 3034-53-5
EINECS: 221-229-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR