2-Bromo-5-fluorobenzaldehyde
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
AS20385G100
AS20385G25
ACTIAS20385G100EA
0
GBP
ACTIAS20385G100
ACTIAS20385G25
2-Bromo-5-fluorobenzaldehyde
2-Bromo-5-fluorobenzaldehyde
Formula:
C₇H₄BrFO MW: 203.01 g/mol |
MDL Number:
MFCD00142872 CAS Number: 94569-84-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...