2-(4-Fluorophenyl)ethanol

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 4-Fluorophenethyl alcohol

AS24775G100 AS24775G25
ACTIAS24775G100EA 166 GBP
ACTIAS24775G100 ACTIAS24775G25
2-(4-Fluorophenyl)ethanol
2-(4-Fluorophenyl)ethanol
Formula: C₈H₉FO
MW: 140.16 g/mol
Boiling Pt: 59…61 °C (0.5 mmHg)
Density: 1,137 g/cm³ (20 °C)
MDL Number: MFCD00002898
CAS Number: 7589-27-7
EINECS: 231-499-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR