1-Cyclohexylpiperazine

Supplier: ACTIVATE SCIENTIFIC GMBH
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

ACTIAS20290G5EA 0 GBP
ACTIAS20290G5 ACTIAS20290G25
1-Cyclohexylpiperazine
1-Cyclohexylpiperazine
Formula: C₁₀H₂₀N₂
MW: 168.28 g/mol
Boiling Pt: 256 °C (1013 hPa)
Melting Pt: 28…32 °C
MDL Number: MFCD00044809
CAS Number: 17766-28-8
EINECS: 241-750-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR