4-Fluoro-m-toluonitrile

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS22529G100 AS22529G25 AS22529G5
ACTIAS22529G100EA 0 GBP
ACTIAS22529G100 ACTIAS22529G25 ACTIAS22529G5
4-Fluoro-m-toluonitrile
4-Fluoro-m-toluonitrile
Formula: C₈H₆FN
MW: 135.14 g/mol
MDL Number: MFCD03788510
CAS Number: 185147-08-4
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR