4-Fluoro-o-toluonitrile

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS21335G100 AS21335G25
ACTIAS21335G100EA 106 GBP
ACTIAS21335G100 ACTIAS21335G25
4-Fluoro-o-toluonitrile
4-Fluoro-o-toluonitrile
Formula: C₈H₆FN
MW: 135.14 g/mol
MDL Number: MFCD03095106
CAS Number: 147754-12-9
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR