2-(4-Chlorophenyl)ethanol

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 4-Chlorophenethyl alcohol

AS25289G100 AS25289G25
ACTIAS25289G100EA 0 GBP
ACTIAS25289G100 ACTIAS25289G25
2-(4-Chlorophenyl)ethanol
2-(4-Chlorophenyl)ethanol
Formula: C₈H₉ClO
MW: 156.61 g/mol
Boiling Pt: 259 °C (1013 hPa)
Density: 1.189 g/cm³ (25 °C)
MDL Number: MFCD00002899
CAS Number: 1875-88-3
EINECS: 217-506-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR