2-(3-Chlorophenyl)ethanol

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 3-Chlorophenethyl alcohol

AS21645G1 AS21645G25 AS21645G5
ACTIAS21645G1EA 14 GBP
ACTIAS21645G1 ACTIAS21645G25 ACTIAS21645G5
2-(3-Chlorophenyl)ethanol
2-(3-Chlorophenyl)ethanol
Formula: C₈H₉ClO
MW: 156.61 g/mol
Boiling Pt: 257 °C (1013 hPa)
Density: 1.189 g/cm³ (20 °C)
MDL Number: MFCD00002892
CAS Number: 5182-44-5
EINECS: 225-964-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR