(3-Chlorophenoxy)acetic acid

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 3-CPA, 3-Chlorophenoxyacetic acid

AS22916G100 AS22916G25
ACTIAS22916G100EA 198 GBP
ACTIAS22916G100 ACTIAS22916G25
(3-Chlorophenoxy)acetic acid
(3-Chlorophenoxy)acetic acid
Formula: (C₆H₄Cl)OCH₂COOH
MW: 186.59 g/mol
Boiling Pt: 308 °C (1013 hPa)
Melting Pt: 110 °C
MDL Number: MFCD00015850
CAS Number: 588-32-9
EINECS: 209-616-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR