(S)-(+)-1-Phenyl-1,2-ethanediol

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: (S)-(+)-Phenyl-1,2-ethanediol, (S)-1-Phenylethane-1,2-diol, (S)-(+)-Phenylethane-1,2-diol, (S)-(+)-α,β-Dihydroxyethylbenzene

AS25093G25 AS25093G5
ACTIAS25093G25EA 154 GBP
ACTIAS25093G25 ACTIAS25093G5
(S)-(+)-1-Phenyl-1,2-ethanediol
(S)-(+)-1-Phenyl-1,2-ethanediol
Formula: C₈H₁₀O₂
MW: 138.17 g/mol
Boiling Pt: 272 °C (1013 hPa)
Melting Pt: 67 °C
MDL Number: MFCD00066256
CAS Number: 25779-13-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR