L-Homoserine
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
H-HomoSer-OH, H-HoSer-OH, H-Hser-OH, (S)-(-)-Homoserine, L(-)-Homoserine
AS22074G25
AS22074G5
ACTIAS22074G25EA
83
GBP
ACTIAS22074G25
ACTIAS22074G5
L-Homoserine
L-Homoserine
Formula:
C₄H₉NO₃ MW: 119.12 g/mol Melting Pt: 203 °C |
MDL Number:
MFCD00063090 CAS Number: 672-15-1 EINECS: 211-590-6 Merck Index: 13,04760 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...