2,6-Dichloro-α-methylbenzyl alcohol

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS21377G25 AS21377G5
ACTIAS21377G25EA 241 GBP
ACTIAS21377G25 ACTIAS21377G5
2,6-Dichloro-α-methylbenzyl alcohol
2,6-Dichloro-α-methylbenzyl alcohol
Formula: C₈H₈Cl₂O
MW: 191.06 g/mol
MDL Number: MFCD00016853
CAS Number: 53066-19-6
EINECS: 258-340-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR