Tetrahydro-4H-pyran-4-one

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Dihydro-2H-pyran-4(3H)-one, Tetrahydro-α-pyran-4-one, Tetrahydro-2H-pyran-4-one, Tetrahydro-γ-pyran-4-one

AS7034G100 AS7034G25
ACTIAS7034G100EA 0 GBP
ACTIAS7034G100 ACTIAS7034G25
Tetrahydro-4H-pyran-4-one
Tetrahydro-4H-pyran-4-one
Formula: C₅H₈O₂
MW: 100.12 g/mol
Boiling Pt: 166 °C (1013 hPa)
Density: 1.08 g/cm³ (20 °C)
MDL Number: MFCD00006581
CAS Number: 29943-42-8
EINECS: 249-967-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR