2,7-Dibromo-9-fluorenone

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2,7-Dibromo-9H-fluoren-9-one

AS21479G100 AS21479G25
ACTIAS21479G100EA 113 GBP
ACTIAS21479G100 ACTIAS21479G25
2,7-Dibromo-9-fluorenone
2,7-Dibromo-9-fluorenone
Formula: C₁₃H₆Br₂O
MW: 338 g/mol
MDL Number: MFCD00010790
CAS Number: 14348-75-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR