2-Methylbenzyl alcohol

Supplier: ACTIVATE SCIENTIFIC GMBH
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

ACTIAS24079G100EA 129 GBP
ACTIAS24079G100
2-Methylbenzyl alcohol
2-Methylbenzyl alcohol
Formula: C₈H₁₀O
MW: 122.17 g/mol
Boiling Pt: 224 °C (1013 hPa)
Melting Pt: 31…35 °C
Density: 1.08 g/cm³ (20 °C)
Flash Pt: 104 °C
MDL Number: MFCD00004622
CAS Number: 89-95-2
EINECS: 201-954-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR