(3-Fluorophenyl)acetonitrile
Supplier: ACTIVATE SCIENTIFIC GMBH
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
2-(3-Fluorophenyl)acetonitrile
, 3-Fluorobenzyl cyanide
ACTIAS24934G100EA
212
GBP
ACTIAS24934G100
ACTIAS24934G25
(3-Fluorophenyl)acetonitrile
(3-Fluorophenyl)acetonitrile
Formula:
C₈H₆FN MW: 135.14 g/mol Boiling Pt: 113 °C (1013 hPa) Density: 1.14058 g/cm³ (15 °C) |
MDL Number:
MFCD00001907 CAS Number: 501-00-8 EINECS: 207-918-2 UN: 3276 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...