1-Nitro-4-(trifluoromethoxy)benzene

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 4-(Trifluoromethoxy)nitrobenzene

AS24740G100 AS24740G25
ACTIAS24740G100EA 87 GBP
ACTIAS24740G100 ACTIAS24740G25
1-Nitro-4-(trifluoromethoxy)benzene
1-Nitro-4-(trifluoromethoxy)benzene
Formula: C₇H₄F₃NO₃
MW: 207.11 g/mol
MDL Number: MFCD00040302
CAS Number: 713-65-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR