3-Fluoro-4-hydroxybenzoic acid

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS25001G100 AS25001G25
ACTIAS25001G100EA 224 GBP
ACTIAS25001G100 ACTIAS25001G25
3-Fluoro-4-hydroxybenzoic acid
3-Fluoro-4-hydroxybenzoic acid
Formula: C₇H₅FO₃
MW: 156.11 g/mol
MDL Number: MFCD00143161
CAS Number: 350-29-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR