Methyl-4-bromobenzoate

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Methyl 4-bromobenzoate, 4-Bromobenzoic acid methyl ester

AS20132G100 AS20132G25
ACTIAS20132G100EA 0 GBP
ACTIAS20132G100 ACTIAS20132G25
Methyl-4-bromobenzoate
Methyl 4-bromobenzoate
Formula: C₈H₇BrO₂
MW: 215.05 g/mol
Boiling Pt: 246 °C (1013 hPa)
Melting Pt: 78…81 °C
Density: 1.689 g/cm³ (20 °C)
MDL Number: MFCD00013531
CAS Number: 619-42-1
EINECS: 210-596-6
Merck Index: 13,01392

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR