(R)-(-)-2-Amino-3-methyl-1-butanol

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: D(-)-2-Amino-3-methyl-1-butanol, (R)-(-)-2-Amino-3-methylbutanol, D-Valinol, D(-)-Valinol

AS22326G25 AS22326G5
ACTIAS22326G25EA 0 GBP
ACTIAS22326G25 ACTIAS22326G5
(R)-(-)-2-Amino-3-methyl-1-butanol
(R)-(-)-2-Amino-3-methyl-1-butanol
Formula: C₅H₁₃NO
MW: 103.16 g/mol
Density: 0.93 g/cm³ (20 °C)
MDL Number: MFCD00064297
CAS Number: 4276-09-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR