(R)-(-)-2-Amino-1-propanol

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: D-Alaninol, D(-)-2-Amino-1-propanol, D(-)-Alaninol, (R)-2-Aminopropan-1-ol

AS20486G100 AS20486G25
ACTIAS20486G100EA 0 GBP
ACTIAS20486G100 ACTIAS20486G25
(R)-(-)-2-Amino-1-propanol
(R)-(-)-2-Amino-1-propanol
Formula: C₃H₉NO
MW: 75.11 g/mol
Boiling Pt: 173 °C (1013 hPa)
Density: 0.96 g/cm³ (20 °C)
MDL Number: MFCD00064413
CAS Number: 35320-23-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR