6-Aminoindole

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS21643G1 AS21643G5
ACTIAS21643G1EA 43 GBP
ACTIAS21643G1 ACTIAS21643G5
6-Aminoindole
6-Aminoindole
Formula: C₈H₈N₂
MW: 132.16 g/mol
Boiling Pt: 161…166 °C (2,3 mmHg)
Melting Pt: 65…66 °C
Storage Temperature: Freezer
MDL Number: MFCD02093959
CAS Number: 5318-27-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR