4-Chloro-3-fluorobenzoic acid

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

AS21639G25 AS21639G5
ACTIAS21639G25EA 47 GBP
ACTIAS21639G25 ACTIAS21639G5
4-Chloro-3-fluorobenzoic acid
4-Chloro-3-fluorobenzoic acid
Formula: C₇H₄ClFO₂
MW: 174.56 g/mol
MDL Number: MFCD00143290
CAS Number: 403-17-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR