5-Methylisatin
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
5-Methylindoline-2,3-dione, 5-Methyl-1H-indole-2,3-dione
AS22643G100
AS22643G25
ACTIAS22643G100EA
81
GBP
ACTIAS22643G100
ACTIAS22643G25
5-Methylisatin
5-Methylisatin
Formula:
C₉H₇NO₂ MW: 161.16 g/mol |
MDL Number:
MFCD00005721 CAS Number: 608-05-9 EINECS: 210-152-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...