3-Fluoro-2-hydroxypyridine

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS20313G25 AS20313G5
ACTIAS20313G25EA 138 GBP
ACTIAS20313G25 ACTIAS20313G5
3-Fluoro-2-hydroxypyridine
3-Fluoro-2-hydroxypyridine
Formula: C₅H₄FNO
MW: 113.09 g/mol
MDL Number: MFCD03095042
CAS Number: 1547-29-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR