Pyridine-3,5-dicarboxylic acid

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: Dinicotinic acid, 3,5-Pyridinedicarboxylic acid

AS20655G100 AS20655G25
ACTIAS20655G100EA 89 GBP
ACTIAS20655G100 ACTIAS20655G25
Pyridine-3,5-dicarboxylic acid
Pyridine-3,5-dicarboxylic acid
Formula: C₇H₅NO₄
MW: 167.12 g/mol
Boiling Pt: 357 °C (1013 hPa)
Melting Pt: 320…325 °C
Storage Temperature: Ambient
MDL Number: MFCD00006393
CAS Number: 499-81-0
EINECS: 207-893-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR