4-Bromo-2,6-difluoroaniline

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS23134G100 AS23134G25
ACTIAS23134G100EA 64 GBP
ACTIAS23134G100 ACTIAS23134G25
4-Bromo-2,6-difluoroaniline
4-Bromo-2,6-difluoroaniline
Formula: C₆H₄BrF₂N
MW: 208.01 g/mol
MDL Number: MFCD00013389
CAS Number: 67567-26-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR