3,5-Dichlorophenyl isocyanate

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 3,5-Dichlorophenylisocyantae

AS20148G100 AS20148G25
ACTIAS20148G100EA 0 GBP
ACTIAS20148G100 ACTIAS20148G25
3,5-Dichlorophenyl isocyanate
3,5-Dichlorophenyl isocyanate
Formula: C₇H₃Cl₂NO
MW: 188.01 g/mol
Boiling Pt: 264 °C (1013 hPa)
Melting Pt: 32…34 °C
Density: 1.38 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00013859
CAS Number: 34893-92-0
EINECS: 252-276-9
UN: 2250
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR