(S)-(+)-1-Aminopropan-2-ol

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

Synonyms: (S)-1-Aminopropan-2-ol, (S)-(+)-Isopropanolamine, (S)-(+)-1-Amino-2-propanol, (+)-1-Aminopropan-2-ol, (S)-Isopropanolamine, (+)-Isopropanolamine

AS20465G100 AS20465G25 AS20465G5
ACTIAS20465G100EA 0 GBP
ACTIAS20465G100 ACTIAS20465G25 ACTIAS20465G5
(S)-(+)-1-Aminopropan-2-ol
(S)-(+)-1-Aminopropan-2-ol
Formula: C₃H₉NO
MW: 75.11 g/mol
Boiling Pt: 160 °C (1013 hPa)
Melting Pt: –2 °C
Density: 0.96 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064429
CAS Number: 2799-17-9
EINECS: 220-533-4
UN: 3259
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR