(R)-(-)-1-Aminopropan-2-ol
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
Synonyms:
(R)-Isopropanolamine, (R)-(-)-1-Amino-2-propanol, (R)-1-Aminopropan-2-ol, (-)-1-Aminopropan-2-ol, (R)-(-)-Isopropanolamine, (-)-Isopropanolamine, (R)-1-Amino-2-propanol
AS20464G100
AS20464G25
AS20464G5
ACTIAS20464G100EA
0
GBP
ACTIAS20464G100
ACTIAS20464G25
ACTIAS20464G5
(R)-(-)-1-Aminopropan-2-ol
(R)-(-)-1-Aminopropan-2-ol
Formula:
C₃H₉NO MW: 75.11 g/mol Boiling Pt: 160 °C (1013 hPa) Melting Pt: 24…26 °C Density: 0.95 g/cm³ (20 °C) Storage Temperature: Refrigerator |
MDL Number:
MFCD00064428 CAS Number: 2799-16-8 EINECS: 220-532-9 UN: 3259 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...