4-Aminopyrimidine

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

AS22453G1 AS22453G5
ACTIAS22453G1EA 0 GBP
ACTIAS22453G1 ACTIAS22453G5
4-Aminopyrimidine
4-Aminopyrimidine
Formula: C₄H₅N₃
MW: 95.1 g/mol
Melting Pt: 154…156 °C
MDL Number: MFCD00006112
CAS Number: 591-54-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR