2-Chloro-4-fluoropyridine

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS22145G25 AS22145G5
ACTIAS22145G25EA 0 GBP
ACTIAS22145G25 ACTIAS22145G5
2-Chloro-4-fluoropyridine
2-Chloro-4-fluoropyridine
Formula: C₅H₃ClFN
MDL Number: MFCD04039345
CAS Number: 34941-91-8
UN: 2929
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR