(2,3-Dichlorophenyl)acetic acid

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2,3-Dichlorophenylacetic acid

AS20899G100 AS20899G25
ACTIAS20899G100EA 146 GBP
ACTIAS20899G100 ACTIAS20899G25
(2,3-Dichlorophenyl)acetic acid
(2,3-Dichlorophenyl)acetic acid
Formula: (C₆H₃Cl₂)CH₂COOH
MW: 205.04 g/mol
MDL Number: MFCD01861393
CAS Number: 10236-60-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR