2,4,6-Trichloropyridine

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS22094G1 AS22094G25 AS22094G5
ACTIAS22094G1EA 0 GBP
ACTIAS22094G1 ACTIAS22094G25 ACTIAS22094G5
2,4,6-Trichloropyridine
2,4,6-Trichloropyridine
Formula: C₅H₂Cl₃N
MW: 182.44 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00971928
CAS Number: 16063-69-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR