2-Bromo-3-fluoropyridine
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
AS50690G1
AS50690G5
ACTIAS50690G1EA
0
GBP
ACTIAS50690G1
ACTIAS50690G5
2-Bromo-3-fluoropyridine
2-Bromo-3-fluoropyridine
Formula:
C₅H₃BrFN |
MDL Number:
MFCD04114128 CAS Number: 40273-45-8 UN: 2810 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...