3-Formylphenylboronic acid

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Dihydroxy(3-formylphenyl)borane, 3-Boronobenzaldehyde, (3-Formylphenyl)metaboric acid, 3-Formylbenzeneboronic acid, (3-Formylphenyl)boronic acid

AS50707G100 AS50707G5
ACTIAS50707G100EA 0 GBP
ACTIAS50707G100 ACTIAS50707G5
3-Formylphenylboronic acid
3-Formylphenylboronic acid
Formula: C₇H₇BO₃
MW: 149.94 g/mol
MDL Number: MFCD00161356
CAS Number: 87199-16-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR