Potassium peroxodisulphate ≥99.0% ACS
Supplier: Thermo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
Synonyms:
Dipotassium peroxodisulphate
, Peroxydisulphuric acid dipotassium salt
, Potassium persulphate
, di-Potassium peroxodisulphate
46939.30EA
41.4
GBP
46939.30
46939.A1
Potassium peroxodisulphate ≥99.0% ACS
Potassium peroxodisulphate
Formula:
K₂S₂O₈ MW: 270.32 g/mol Melting Pt: 100 °C (dec.) Density: 2.477 Storage Temperature: Ambient |
MDL Number:
MFCD00011386 CAS Number: 7727-21-1 EINECS: 231-781-8 UN: 1492 ADR: 5.1,III Merck Index: 14,7656 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...