4,7-Dichloroisatin

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: 4,7-Dichloroindoline-2,3-dione, 4,7-Dichloro-1H-indole-2,3-dione

AS24430G25 AS24430G5
ACTIAS24430G25EA 180 GBP
ACTIAS24430G25 ACTIAS24430G5
4,7-Dichloroisatin
4,7-Dichloroisatin
Formula: C₈H₃Cl₂NO₂
MW: 216.02 g/mol
MDL Number: MFCD00047214
CAS Number: 18711-13-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR